Human recombinant Cytochrome b5A (from E. coli)
Fournisseur: ProSci Inc.
91-197
PRSI91-197EA 761 EUR InStock
PRSI91-197
Human recombinant Cytochrome b5A (from E. coli)
Protéines et Peptides
Cytochrome b5 (CYB5A) is a membrane bound hemoprotein which function as an electron carrier for several membrane bound oxygenases. CYB5A contains one cytochrome b5 heme-binding domain and has two isoforms produced by alternative splicing. Isoform 1 is a sngle-pass membrane protein. Isoform 2 is located in cytoplasm. The defects in CYB5A can result in type IV hereditary methemoglobinemia.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité