Human Recombinant EIF1B (from E. coli)
Fournisseur: ProSci Inc.
91-200
PRSI91-200EA 761 EUR InStock
PRSI91-200
Human Recombinant EIF1B (from E. coli)
Protéines et Peptides
Eukaryotic Translation Initiation Factor 1B (EIF1B) is an element of a complex involved in recognition of the initiator codon during the scanning process. Translation is also initiated by the function of EIF1B in regulating the activity of ribosomal subunits 43S, 48S and 40S. EIF1B enables 43S ribosomal complexes to distinguish between cognate and near-cognate initiation codons, perceiving the nucleotide content of initiation codons.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité