Human Recombinant GAS7 (from E. coli)
Fournisseur: ProSci Inc.
91-205
PRSI91-205EA 884 EUR InStock
PRSI91-205
Human Recombinant GAS7 (from E. coli)
Protéines et Peptides
Growth Arrest-Specific Protein 7 (GAS7) is expressed primarily in terminalaly differentiated brain cells and predominantly in mature cerebellar Purkinje neurons. GAS7 may play a role in neuronal development by promoting maturation and morphological differentiation of cerebellar neurons. Inhibition of GAS7 production in terminally differentiating cultures of embryonic murine cerebullum impedes neurite outgrowth. The hyper-expression of GAS7 may play an major role in the initiation and development of huaman osteosarcoma.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité