Human recombinant GDNF (from E. coli)
Fournisseur: ProSci Inc.
91-206
PRSI91-206EA 852 EUR InStock
PRSI91-206
Human recombinant GDNF (from E. coli)
Protéines et Peptides
Glial Cell Line-Derived Neurotrophic Factor (GDNF) is a disulfide-linked homodimeric glycoprotein that belongs to the TGF- beta superfamily. It has been shown to promote the survival of various neuronal subpopulations in both the central as well as the peripheral nervous systems at different stages of their development. Human GDNF cDNA encodes a 211 amino acid residue prepropeptide that is processed to yield a dimeric protein. Mature human GDNF was predicted to contain two 134 amino acid residue subunits. Cells known to express GDNF include Sertoli cells, type 1 astrocytes, Schwann cells, neurons, pinealocytes and skeletal muscle cells. Mutations in this gene may be associated with Hirschsprung disease.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité