Human Recombinant Hemoglobin Zeta (from E. coli)
Fournisseur: ProSci Inc.
91-211
PRSI91-211EA 789 EUR InStock
PRSI91-211
Human Recombinant Hemoglobin Zeta (from E. coli)
Protéines et Peptides
Hemoglobin Subunit Zeta (HBZ) is a member of the Globin family. The zeta chain is an alpha-type chain of mammalian embryonic Hemoglobin that is synthesized primarily in the yolk sac of the early embryo, while alpha-globin is produced throughout fetal growth and adult life. The HBZ gene consists of five functional genes and two pseudogenes, the order of genes is 5-zeta-pseudozeta-mu-pseudoalpha-1-alpha-2-alpha-1-theta-1-3.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité