Human Recombinant HLP2 (from E. coli)
Fournisseur: ProSci Inc.
91-212
PRSI91-212EA 884 EUR InStock
PRSI91-212
Human Recombinant HLP2 (from E. coli)
Protéines et Peptides
Hippocalcin-Like Protein 1 (HPCAL1) is a neuron-specific calcium-binding member of the recoverin family which found in the retina and brain. HPCAL1 contains four EF-hand domains and it is highly similar to human hippocalcin protein. HPCAL1 is involved in the calcium-dependent regulation of rhodopsin phosphorylation. In addition, it may be of relevance for neuronal signalling in the central nervous system.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité