Human recombinant Myelin P2 (from E. coli)
Fournisseur: ProSci Inc.
91-218
PRSI91-218EA 852 EUR InStock
PRSI91-218
Human recombinant Myelin P2 (from E. coli)
Protéines et Peptides
Myelin P2 Protein (PMP2) is a cytoplasmic protein which belongs to the Fatty-acid binding protein (FABP) family of calycin superfamily. PMP2 is a small, basic, and cytoplasmic lipid binding protein of peripheral myelin. PMP2 is found in peripheral nerve myelin and spinal cord myelin, the oligodendrocytes and Schwann cells, respectively. PMP2 may be involved in lipid transport protein in Schwann cells. It may decrease the inhibitory effect of T suppressors in the culture of immune lymph node cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité