Human recombinant Neurocalcin delta (from E. coli)
Fournisseur: ProSci Inc.
91-219
PRSI91-219EA 884 EUR InStock
PRSI91-219
Human recombinant Neurocalcin delta (from E. coli)
Protéines et Peptides
Neurocalcin-delta (NCALD) is a neuronal calcium-binding protein that belongs to the neuronal calcium sensor (NCS) family. It expressed in mammalian brains. NCALD contains an N-terminal myristoylation signal and four EF-hand calcium binding loops. The protein possesses a Ca2+ /myristoyl switch. It is cytosolic at resting calcium levels. However, elevated intracellular calcium levels induce a conformational change which exposes the myristoyl group, resulting in protein association with membranes and partial co-localization with the perinuclear trans-golgi network. NCALD protein is thought to be a regulator of G protein-coupled receptor signal transduction.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité