Human Recombinant NHP2L1 (from E. coli)
Fournisseur: ProSci Inc.
91-220
PRSI91-220EA 852 EUR InStock
PRSI91-220
Human Recombinant NHP2L1 (from E. coli)
Protéines et Peptides
NHP2-Like Protein 1 (NHP2L1) is a member of the ribosomal protein L7Ae family. NHP2L1 proteinis limited to the nucleus, primarily focused in the dense fibrillar component of the nucleolus. NHP2L1 has been shown to interact with RAD17and PRPF31. The protein undergoes a conformational change upon RNA-binding. NHP2L1 binds to the 5-stem-loop of U4 snRNA and may play a role in the late stage of spliceosome assembly, prior to step I of splicing catalysis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité