Human recombinant p18-INK4c (from E. coli)
Fournisseur: ProSci Inc.
91-223
PRSI91-223EA 852 EUR InStock
PRSI91-223
Human recombinant p18-INK4c (from E. coli)
Protéines et Peptides
Cyclin-Dependent Kinase 4 Inhibitor C (CDKN2C) is a member of the INK4 family of cyclin dependent kinase inhibitors. CDKN2C contains 4 ANK repeats and interacts with CDK4 or CDK6. Highest levels of CDKN2C can be found in skeletal muscle, pancreas, and heart. CDKN2C inhibits cell growth and proliferation with a correlated dependence on endogenous retinoblastoma protein RB and prevent the activation of the CDK kinases. Studies have been shown the roles of CDKN2C gene in regulating spermatogenesis, as well as in suppressing tumorigenesis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité