Human recombinant PAF-AHB (from E. coli)
Fournisseur: ProSci Inc.
91-224
PRSI91-224EA 884 EUR InStock
PRSI91-224
Human recombinant PAF-AHB (from E. coli)
Protéines et Peptides
Platelet-Activating Factor Acetylhydrolase IB Subunit beta (PAFAHB) is a cytoplasmic hydrolase. PAFAHB is a member of the GDSL lipolytic enzyme family. It also belongs to Platelet-activating factor acetylhydrolase IB beta/gamma subunits subfamily. Cytosolic PAF-AH IB is formed of three subunits of 45 kDa (alpha), 30 kDa (beta) and 29 kDa (gamma), PAFAHB is a catalytic subunit. PAFAHB inactivates PAF by removing the acetyl group at the sn-2 position.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité