Human recombinant PHS (from E. coli)
Fournisseur: ProSci Inc.
91-226
PRSI91-226EA 884 EUR InStock
PRSI91-226
Human recombinant PHS (from E. coli)
Protéines et Peptides
Pterin-4- alpha-Carbinolamine Dehydratase (PCBD1) is the founding member of the Pterin-4- alpha-Carbinolamine Dehydratase Family. PCBD1 is involved in Tetrahydrobiopterin biosynthesis. It seems to prevent the formation of 7-Pterins and accelerate the formation of Quinonoid-BH2. Furthermore, PCBD1 regulates the homodimerization of the transcription factor Hepatocyte Nuclear Factor 1 (HNF1) and enhances its transcriptional activity. Defects in PCBD1 are the cause of BH4-Deficient Hyperphenylalaninemia Type D (HPABH4D). HPABH4D is characterized by the excretion of 7-substituted Pterins in the urine of affected patients.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité