Human recombinant PKI-beta (from E. coli)
Fournisseur: ProSci Inc.
91-227
PRSI91-227EA 884 EUR InStock
PRSI91-227
Human recombinant PKI-beta (from E. coli)
Protéines et Peptides
cAMP-Dependent Protein Kinase Inhibitor beta (PKI- beta ) is a member of the PKI family. It has been shown that PKI- beta is an extremely potent competitive inhibitor of cAMP-dependent protein kinase activity; this protein interacts with the catalytic subunit of the enzyme after the cAMP-induced dissociation of its regulatory chains.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité