Human recombinant Phosphomannomutase 1 (from E. coli)
Fournisseur: ProSci Inc.
91-229
PRSI91-229EA 884 EUR InStock
PRSI91-229
Human recombinant Phosphomannomutase 1 (from E. coli)
Enzymes
Phosphomannomutase 1 (PMM1) blongs to the eukaryotic PMM family. Phosphomannomutase 1 can catalyses the conversion between D-mannose 6-phosphate and D-mannose 1-phosphate which is a substrate for GDP-mannose synthesis. GDP-mannose is used for synthesis of dolichol-phosphate-mannose which required for a number of critical mannosyl transfer reactions. PMM1 is highly expressed in liver, heart, brain, and pancreas, but lower expression in skeletal muscle. In addition, PMM1 may be responsible for the degradation of glucose-1,6 bisphosphate in ischemic brain.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité