Human recombinant Phosphomannomutase 2 (from E. coli)
Fournisseur: ProSci Inc.
91-230
PRSI91-230EA 789 EUR InStock
PRSI91-230
Human recombinant Phosphomannomutase 2 (from E. coli)
Enzymes
Phosphomannomutase 2 (PMM2) is an enzyme that is a member of the highly variable methyltransferase superfamily. PMM2 is a cytoplasmic protein and catalyses the isomerisaion of mannose 6-phosphate to mannose 1-phosphate.In addition, PMM2 involved in the synthesis of the GDP-mannose and dolichol-phosphate-mannose that required for a number of critical mannosyl transfer reactions. Defects in PMM2 can results in congenital disorder of glycosylation type 1A (CDG1A). Congenital disorders of glycosylation are metabolic deficiencies in glycoprotein biosynthesis that usually cause severe mental and psychomotor retardation.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité