Human Recombinant Heat Shock beta 11 (from E. coli)
Fournisseur: ProSci Inc.
91-231
PRSI91-231EA 884 EUR InStock
PRSI91-231
Human Recombinant Heat Shock beta 11 (from E. coli)
Protéines et Peptides
Heat Shock Protein beta -11 (HSPB11) is a stress-responsive protein that is required to deal with proteotoxic stresses. HSPB11 is composed of an IFT complex B composed of IFT88, IFT57, TRAF3IP1, IFT52, IFT27, HSPB11 and IFT20 and is detected in placenta. HSPB11 has beeb shown to form oligomeric complexes and prevent the aggregation of in vitro denaturated aldolase and glyceraldehyde-3-phosphate dehydrogenase in accordance with the chaperone model of HSPB1 and HSPB5. HSPB11 overexpression protected against etoposide-induced cell death that correlated with a decreased release of mitochondrial Cytochrome C into the cytosol. Inhibition of HSP90 function completely abrogated the protective effect of HSPB11. This data suggests that at least in the case of HSPB11, interaction with other chaperone machines besides HSPA1A may contribute to functional specificity and cellular functioning.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité