Human recombinant PPC-DC (from E. coli)
Fournisseur: ProSci Inc.
91-232
PRSI91-232EA 884 EUR InStock
PRSI91-232
Human recombinant PPC-DC (from E. coli)
Protéines et Peptides
Phosphopantothenoylcysteine Decarboxylase (PPC-DC) is an essential enzyme in the biosynthesis ofCoenzyme A and catalyzes the decarboxylation of PPC to Phosphopantetheine. PPC-DC catalyzes the decarboxylation of the Cysteine moiety of 4-Phosphopantothenoylcysteine (PPC) to form 4-Phosphopantetheine (PPantSH), this reaction forms part of the biosynthesis of Coenzyme A. The enzyme is a member of the larger family of Cysteine Decarboxylases including the Lantibiotic-Biosynthesizing enzymes EpiD and MrsD, all of which use a tightly bound Flavin cofactor to oxidize the Thiol moiety of the substrate to a Thioaldehyde.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité