Human recombinant protein CutA (from E. coli)
Fournisseur: ProSci Inc.
91-235
PRSI91-235EA 852 EUR InStock
PRSI91-235
Human recombinant protein CutA (from E. coli)
Protéines et Peptides
Protein CutA (CUTA) posseses a signal peptide and is widely expressed in brain. CUTA mayforms part of a complex of membrane proteins attached to acetylcholinesterase (AChE). CUTA takes part in cellular tolerance to a broad range of divalent cations other than copper. Alternate transcriptional splice variants, both protein-coding and non-protein-coding, have been found.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité