Human recombinant S100 calcium binding P (from E. coli)
Fournisseur: ProSci Inc.
91-238
PRSI91-238EA 789 EUR InStock
PRSI91-238
Human recombinant S100 calcium binding P (from E. coli)
Protéines et Peptides
Protein S100-P (S100P) belongs to the S100 family of Calcium-Binding Proteins. S100P is 95 amino acids in length and it contains 2 EF-Hand Calcium-Binding Motifs. The EF-Hand Motif binds calcium, which likely alters molecular conformation. S100 proteins are localized in the cytoplasm and/or nucleus of a wide range of cells and it is involved in the regulation of a number of cellular processes such as cell cycle progression and differentiation. In addition to binding Ca, S100P also binds Zn and Mg, and may play a role in the etiology of prostate cancer.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité