Human recombinant SCO1 (from E. coli)
Fournisseur: ProSci Inc.
91-239
PRSI91-239EA 852 EUR InStock
PRSI91-239
Human recombinant SCO1 (from E. coli)
Protéines et Peptides
Protein SCO1 Homolog, Mitochondrial (SCO1) is a member of the SCO1/2 family. SCO1 has a homodimer structure. SCO1 is located in mitochondrion and is highly expressed in muscle, heart, and brain. It is characterized by high rates of Oxidative Phosphorylation (OxPhos). SCO1 is thought to play a important role in cellular copper homeostasis, mitochondrial redox signaling and insertion of copper into the active site of COX. The defects of SCO1 can result in Mitochondrial Complex IV Deficiency (MT-C4D). A disorder of the mitochondrial respiratory chain has heterogeneous clinical manifestations, ranging from isolated myopathy to severe multisystem disease affecting several tissues and organs.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité