Human recombinant CLIC2 (from E. coli)
Fournisseur: ProSci Inc.
91-246
PRSI91-246EA 884 EUR InStock
PRSI91-246
Human recombinant CLIC2 (from E. coli)
Protéines et Peptides
Chloride Intracellular Channel Protein 2 (CLIC2) is a critical component of all living cells; it regulatescellular traffic of Chloride ion and it can be inserted into membranes anf form chloride ion channels. Membrane insertion seems to be redox-regulated and may occur only under oxydizing conditions, channel activity depends on the pH. CLIC2 is involved in regulating membrane potential and organic solute transport. CLIC2 modulates the activity of RYR2 and inhibits Calcium influx. CLIC2 can be detected in the adult brain, liver, lung, heart, stomach, spleen and testis. It is expressed in fetal liver and adult skeletal muscle. CLIC2 is a potential candidate for one of many diseases linked to Xq28.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité