Human recombinant ZW10 interactor (fromE. coli)
Fournisseur: ProSci Inc.
91-247
PRSI91-247EA 761 EUR InStock
PRSI91-247
Human recombinant ZW10 interactor (fromE. coli)
Protéines et Peptides
ZW10 Interactor is localized to the kinetochores from late Prophase to Anaphase and has a uniform distribution in the cytoplasm of Interphase cells. ZWINT interacts ZW10, MIS12 and NDC80 subunit of the NDC80 complex specifically during mitosis. ZWINT is a part of the MIS12 complex which is required for kinetochore formation and spindle checkpoint activity. In addition, ZWINT is required to target ZW10 to the kinetochore at prometaphase.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité