Human Recombinant GABARAPL1 (from E. coli)
Fournisseur: ProSci Inc.
91-249
PRSI91-249EA 852 EUR InStock
PRSI91-249
Human Recombinant GABARAPL1 (from E. coli)
Protéines et Peptides
Gamma-Aminobutyric Acid Receptor-Associated Protein-Like 1 (GABARAPL1) is a cytoplasmic protein that belongs to the MAP1 LC3 family. GABARAPL1 is expressed at very high levels in the brain, heart, peripheral blood leukocytes, liver, kidney, placenta, and skeletal muscle. It can interact with GABRG2, OPRK1 and beta -Tubulin. GABARAPL1 increases cell-surface expression of kappa-type opioid receptor through facilitating anterograde intracellular trafficking of the receptor.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité