Human recombinant DCN1-like 1 (from E. coli)
Fournisseur: ProSci Inc.
91-251
PRSI91-251EA 852 EUR InStock
PRSI91-251
Human recombinant DCN1-like 1 (from E. coli)
Protéines et Peptides
DCN1-Like Protein 1 is a protein containing 1 DCUN1 domain and 1 UBA-like domain. DCN1-Like Protein 1 contains part of an E3 Ubiquitin Ligase Complex for Neddylation. It is required for Neddylation of Cullin components of E3 Cullin-RING Ubiquitin Ligase complexes. It enhances the rate of Cullins Neddylation. DCN1-Like Protein 1 recruits the NEDD8-charged E2 Enzyme to the Cullin component. DCUN1D1 is involved in the release of inhibitory effects of CAND1 on the Cullin-RING Ligase E3 complex assembly and activity. It acts also as an oncogene facilitating malignant transformation and carcinogenic progression.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité