Human Recombinant HDHD2 (from E. coli)
Fournisseur: ProSci Inc.
91-254
PRSI91-254EA 947 EUR InStock
PRSI91-254
Human Recombinant HDHD2 (from E. coli)
Protéines et Peptides
Haloacid Dehalogenase-Like Hydrolase Domain-Containing Protein 22 (HDHD2) is a member of the HAD-like hydrolase superfamily. HDHD2 includes L-2-Haloacid Dehalogenase, Epoxide Hydrolases and Phosphatases. There are two active sites in HDHD2 - an L-2-Haloacid Dehalogenase and a Carboxylate group. The L-2-Haloacid Dehalogenase active site catalyzes the hydrolytic dehalogenation of D- and L-2-Haloalkanoic Acids, producing L- and D-2-Hydroxyalkanoic Acids.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité