Human recombinant L-Xylulose Reductase (from E. coli)
Fournisseur: ProSci Inc.
91-257
PRSI91-257EA 789 EUR InStock
PRSI91-257
Human recombinant L-Xylulose Reductase (from E. coli)
Enzymes
L-Xylulose Reductase is an enzyme that belongs to the Short-Chain Dehydrogenases/Reductases (SDR) family. L-Xylulose Reductase is responsible for the metabolism of Xylulose, converting it into Xylitol. L-Xylulose Reductase catalyses the NADPH-dependent reduction of several Pentoses, Tetroses, Trioses, alpha-Dicarbonyl compounds and L-Xylulose. L-Xylulose Reductase participates in the Uronate Cycle of Glucose metabolism. It may play a role in the water absorption and cellular osmoregulation in the proximal renal tubules by producing Xylitol, an osmolyte, thereby preventing osmolytic stress from occurring in the renal tubules.
Fusion-Tag: N-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité