Human recombinant Hydroxyacid Oxidase 1 (from E. coli)
Fournisseur: ProSci Inc.
91-259
PRSI91-259EA 884 EUR InStock
PRSI91-259
Human recombinant Hydroxyacid Oxidase 1 (from E. coli)
Enzymes
Hydroxyacid Oxidase 1 (HAO1) is an enzyme that belongs to the FMN-Dependent alpha-Hydroxy Acid Dehydrogenase family. HAO1 contains 1 FMN Hydroxy Ccid Dehydrogenase domain. HAO1 is expressed primarily in the liver and pancreas. This protein has 2-Hydroxyacid Oxidase activity. Most HAO1 is active on the 2-Carbon substrate Glycolate, but it can also be active on 2-Hydroxy fatty acids, with higher activity towards 2-Hydroxy Palmitate and 2-Hydroxy Octanoate.
  • Fusion-Tag: N-Trx, 6 His tag
Fusion-Tag: N-Trx, 6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité