Human recombinant ZMYND19 (fromE. coli)
Fournisseur: ProSci Inc.
91-260
PRSI91-260EA 852 EUR InStock
PRSI91-260
Human recombinant ZMYND19 (fromE. coli)
Protéines et Peptides
Human Zinc Finger MYND Domain-Containing Protein 19 (ZMYND19) is a protein that contains 1 MYND-Type Zinc Finger. ZMYND19 can be expressed by the brain, testis, placenta, heart, liver, skeletal muscle, kidney, and stomach. ZMYND19 interacts with GPR24/MCH-R1. It binds to the C terminus of Melanin-Concentrating Hormone Receptor-1 and the N Termini of alpha-Tubulin. ZMYND19 may be involved as a regulatory molecule in GPR24/MCH-R1 signaling.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité