Human recombinant ZBP1 (from E. coli)
Fournisseur: ProSci Inc.
91-262
PRSI91-262EA 852 EUR InStock
PRSI91-262
Human recombinant ZBP1 (from E. coli)
Protéines et Peptides
Z-DNA Binding Protein 1 (ZBP1) is a protein with 2 DRADA repeats. ZBP1 is highly expressed in lymphatic tissues including lymph node, leukocytes, tonsil, bone marrow, and spleen. ZBP1 participates in the detection of viral and bacterial DNA from by the host's innate immune system. It plays a role in host defense against tumors and pathogens. ZBP1 Acts as a cytoplasmic DNA sensor which, when activated, induces the recruitment of TBK1 and IRF3 to its C-terminal region and activates the downstream interferon regulatory factor (IRF) and NF-kappa B transcription factors, leading to type-I interferon production. ZBP1-induced NF-kappaB activation probably involves the recruitment of the RHIM containing kinases RIPK1 and RIPK3.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité