Human Recombinant HSF2 (from E. coli)
Fournisseur: ProSci Inc.
91-264
PRSI91-264EA 852 EUR InStock
PRSI91-264
Human Recombinant HSF2 (from E. coli)
Protéines et Peptides
Heat Shock Factor Protein 2 (HSF2) belongs to the HSF family of transcription factors that bind specifically to the heat-shock promoter element and activate transcription. In higher eukaryotes, HSF is unable to bind to the HSE unless the cells are heat shocked. HSF2 is widely expressed in many cells and tissues. HSF2 is located on Cytoplasmic during normal growth. But when it is activited, HSF2 moves to the nucleus.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité