Human Recombinant Galectin 1 (from E. coli)
Fournisseur: ProSci Inc.
91-265
PRSI91-265EA 852 EUR InStock
PRSI91-265
Human Recombinant Galectin 1 (from E. coli)
Protéines et Peptides
Galectin-1 is a member of growing family of evolutionary conserved animal lectins. Galectin-1 is widely expressed in many cells and tissues. Galectins consists of a Galectin domain and two Beta-galactoside binding domains. Galectin-1 can binds LGALS3BP and interacts with CD2, CD3, CD4, CD7, CD43 and CD45. Galectin-1 may act as an autocrine negative growth factor which regulates apoptosis, cell proliferation and cell differentiation. In addition, Galectin-1 plays improtant roles in immunosuppressive and antiinflammatory properties.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité