Human Recombinant MOB4 (from E. coli)
Fournisseur: ProSci Inc.
91-266
PRSI91-266EA 852 EUR InStock
PRSI91-266
Human Recombinant MOB4 (from E. coli)
Protéines et Peptides
MOB-Like Protein Phocein is a member of the MOB1/Phocein Family. MOB-Like Protein Phocein is associated with membranes and the Golgi stacks. It is present in the cytosol, where it behaves as a protein complex. It has been shown that MOB-Like Protein Phocein interacts with DNM1, EPS15 and Nucleoside Diphosphate Kinase. MOB-Like Protein Phocein is the major partner of Striatin Family members and may play a important role in membrane trafficking, specifically in membrane budding reactions.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité