Human recombinant p27 (from E. coli)
Fournisseur: ProSci Inc.
91-268
PRSI91-268EA 761 EUR InStock
PRSI91-268
Human recombinant p27 (from E. coli)
Protéines et Peptides
Cyclin-Dependent Kinase Inhibitor 1B (CDKN1B) is a Kinesin-related motor protein necessary for mitotic spindle assembly and chromosome segregation. CDKN1B is expressed in all tissues with highest levels observed in skeletal muscle. CDKN1B is a potent inhibitor of Cyclin E- and Cyclin A-CDK2 complexes. CDKN1B forms a complex with Cyclin Type D-CDK4 complexes and is involved in the assembly, stability, and modulation of CCND1-CDK4 complex activation. In addition, CDKN1B acts as an inhibitor or an activator of Cyclin Type D-CDK4 complexes depending on its phosphorylation state and stoichometry.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité