Human recombinant UBE2S (from E. coli)
Fournisseur: ProSci Inc.
91-269
PRSI91-269EA 306 EUR InStock
PRSI91-269
Human recombinant UBE2S (from E. coli)
Protéines et Peptides
Ubiquitin-Conjugating Enzyme E2 S (UBE2S) is a member of the Ubiquitin-Conjugating Enzyme family. UBE2S interacts with CDC20, FZR1/CDH1 and VHL. UBE2S can form a thiol ester linkage with Ubiquitin in an Ubiquitin Activating Enzyme-Dependent manner, a characteristic property of Ubiquitin Carrier Proteins. UBE2S acts as an essential factor of the Anaphase Promoting Complex/Cyclosome, a cell cycle-regulated Ubiquitin ligase that controls progression through mitosis. UBE2S is also involved in ubiquitination and subsequent degradation of VHL, resulting in an accumulation of HIF1A.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité