Human Recombinant Histone Chaperone ASF1A (from E. coli)
Fournisseur: ProSci Inc.
91-270
PRSI91-270EA 947 EUR InStock
PRSI91-270
Human Recombinant Histone Chaperone ASF1A (from E. coli)
Protéines et Peptides
Human Histone Chaperone ASF1A (ASF1A) belongs to the H3/H4 family of histone chaperone proteins. ASF1A is ubiquitously expressed in many cells and tissues, interacting with histones H3 and H4. ASF1A cooperates with Chromatin Assembly Factor 1 to promote replication-dependent chromatin assembly and with HIRA to promote replication-independent chromatin assembly. In addition, ASF1A is necessary for the formation of senescence-associated heterochromatin foci (SAHF) and efficient senescence-associated cell cycle exit.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His, N-T7 tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité