Human Recombinant GTF2B (from E. coli)
Fournisseur: ProSci Inc.
91-273
PRSI91-273EA 852 EUR InStock
PRSI91-273
Human Recombinant GTF2B (from E. coli)
Protéines et Peptides
Transcription Initiation Factor IIB (TFIIB) is an essential factor for transcription by RNA Polymerase II. TFIIB localizes to the nucleus where it forms a complex (the DAB complex) with transcription factor IID and IIA. TFIIB plays a role as a bridge between IID, which initially recognizes the promoter sequence, and RNA polymerase II. TFIIB is involved in the selection of transcription start site.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-GST tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité