Human Recombinant HGPRT (from E. coli)
Fournisseur: ProSci Inc.
91-274
PRSI91-274EA 884 EUR InStock
PRSI91-274
Human Recombinant HGPRT (from E. coli)
Protéines et Peptides
Hypoxanthine-Guanine Phosphoribosyltransferase (HGPRT) has an important role in the generation of purine nucleotides through the purine salvage pathway. HPRT1 functions to salvage purines from degraded DNA to renewed purine synthesis, it acts as a catalyst in the reaction between guanine and phosphoribosyl pyrophosphate to form GMP.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité