Human recombinant PDGF-associated (from E. coli)
Fournisseur: ProSci Inc.
91-278
PRSI91-278EA 884 EUR InStock
PRSI91-278
Human recombinant PDGF-associated (from E. coli)
Protéines et Peptides
Human PAP, also known as 28 kDa heat- and acid-stable phosphoprotein, PDGF-associated protein, PDGFA-associated protein 1, PDAP1, HASPP28, is a protein which belongs to the PDAP1 family. The encoded protein in rodents has been shown to bind PDGFA with a low affinity. PDGF-Associated Protein (PAP) is a phosphoprotein that may enhance PDGFA-stimulated cell growth in fibroblasts, but inhibits the mitogenic effect of PDGFB. PDAP1 expression is induced by TNF-alpha, and cells overexpressing PDAP1 show significantly less apoptosis on exposure to TNF-alpha.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité