Human recombinant WARS (from E. coli)
Fournisseur: ProSci Inc.
91-280
PRSI91-280EA 588 EUR InStock
PRSI91-280
Human recombinant WARS (from E. coli)
Protéines et Peptides
There exists two types of tryptophanyl tRNA synthetases, the cytoplasmic form called WARS, the mitochondrial form called WARS2. WARS catalyzes the aminoacylation of tRNA (trp) with tryptophan and is induced by interferon. WARS regulates ERK, Akt, eNOS activation pathway, which are related with angiogenesis, cytoskelatal reorganization and shear stess-reponsive gene expression.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité