Human recombinant BMP Receptor II (from Cells)
Fournisseur: ProSci Inc.
91-283
PRSI91-283EA 409 EUR InStock
PRSI91-283
Human recombinant BMP Receptor II (from Cells)
Protéines et Peptides
Bone Morphogenetic Protein Receptor II (BMPR-II) is a Type II Serine/Threonine Kinase that mediates cellular responses to BMPs. BMPR-II is characterized by lacking of a GS domain, and presence of a C-terminal extension typical of type II receptors. BMPRII binds BMP2, BMP4 and BMP7 weakly in the absence of type I receptor, and the binding can be facilitated by the presence of the type I receptor, including BMPR-IA/Brk1, BMPR-IB, and ActR-I. BMPR-II plays a key role in cell growth. Defects in BMPR-II have been linked to primary pulmonary hypertension. Human and mouse BMPR-II are highly conserved and share 97 % amino acid sequence identity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité