Human recombinant Carbonic Anhydrase 10 (from Cells)
Fournisseur: ProSci Inc.
91-285
PRSI91-285EA 852 EUR InStock
PRSI91-285
Human recombinant Carbonic Anhydrase 10 (from Cells)
Enzymes
Carbonic Anhydrase X (CA10) belongs to CA family of zinc metalloenzymes, which catalyze the reversible hydration of carbon dioxide in various biological processes such as respiration, renal tubular acidification and bone resorption. While CA10 is a secreted protein without Carbonic Anhydrase activity (i.e., the reversible hydration of CO2) due to point mutations in the zinc binding site, it has esterase activity. The human and mouse CA10 are expressed in the brain, indicating that they may play a role in brain development.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité