Human recombinant ICAM2 (from cells)
Fournisseur: ProSci Inc.
91-286
PRSI91-286EA 409 EUR InStock
PRSI91-286
Human recombinant ICAM2 (from cells)
Protéines et Peptides
ICAM-2 is a 55-65 kD transmembrane glycoprotein possessing 2 extracellular Ig domains, a single transmembrane domain, and a short 26-amino acid cytoplasmic domain. ICAM-2 is expressed on most leukocytes, and is strongly expressed on vascular endothelial cells. Interactions of ICAM-2 and the integrin receptors mediate cell adhesion in a wide range of lymphocyte, monocyte, natural killer cell, and granulocytewith other cells, and play important roles in many adhesion-dependent immune and inflammation responses, such as T cell aggregation, NK-cell cytotoxicity and migration, lymphocyte recirculation, etc. Serum levels of ICAM-2 correlated significantly with the inflammatory and course sequences of trichinosis in mice and had a similar relation with blood eosinophilia. So, estimation of ICAM-2 serum levels may prove useful in diagnosis of trichinosis recent infections, and in monitoring the prognosis and response to treatment.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité