Human recombinant KIR2DL4 (from Cells)
Fournisseur: ProSci Inc.
91-290
PRSI91-290EA 549 EUR InStock
PRSI91-290
Human recombinant KIR2DL4 (from Cells)
Protéines et Peptides
Killer cell immunoglobulin-like receptor 2DL4(KIR2DL4) is a Single-pass type I membrane protein and contains 2 Ig-like C2-type (immunoglobulin-like) domains.It belongs to the immunoglobulin superfamily. KIR2DL4 is expressed in all NK cells and some T cells. KIR2DL4 activates the cytotoxicity of NK cells, despite the presence of an immunoreceptor tyrosine-based inhibition motif (ITIM) in its cytoplasmic tail. The ITIM was not necessary for activation of lysis by KIR2DL4. The activation signal of KIR2DL4 was sensitive to inhibition by another ITIM-containing receptor. The activation-deficient mutant of KIR2DL4 inhibited the signal delivered by the activating receptor CD16.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité