Human recombinant TRAILR2 (from cells)
Fournisseur: ProSci Inc.
91-302
PRSI91-302EA 377 EUR InStock
PRSI91-302
Human recombinant TRAILR2 (from cells)
Protéines et Peptides
TNFRSF10B is a member of the TNF-receptor superfamily, and contains an intracellular death domain. This receptor can be activated by tumor necrosis factor-related apoptosis inducing ligand (TNFSF10/TRAIL/APO-2L), and transduces apoptosis signal. The adapter molecule FADD recruits caspase-8 to the activated receptor and is required for the apoptosis mediated by TNFRSF10B. TNFRSF10B is expressed in a number of cell types, and to particularly high levels in lymphocytes and spleen. This single-pass transmembrane protein contains two cysteine-rich repeat units in its extracellular region, followed by a transmembrane segment and a cytoplasmic tail containing a typical “death domain”. TNFRSF10B expression is regulated by the tumor suppressor p53. It is also indicated that the activation of NF-kappa-B can be promoted by TNFRSF10B.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité