Human recombinant CRISP3 (from cells)
Fournisseur: ProSci Inc.
91-306
PRSI91-306EA 549 EUR InStock
PRSI91-306
Human recombinant CRISP3 (from cells)
Protéines et Peptides
Cysteine-rich secretory protein 3 (CRISP-3) is a secreted protein,containing 1 SCP domain and 1 ShKT domain.It is belongs to the CRISP family. CRISP-3 is a glycoprotein that belongs to the family of cysteine-rich secretory proteins (CRISPs) which was originally discovered in human neutrophilic granulocytes. CRISP-3 is also widely distibuted in exocrine glands (salivary glands, pancreas and prostate), eosinophilic granulocytes and to a lower level in epididymis, ovary, thymus and colon. The presence of CRISP-3 in neutrophils, eosinophils and in exocrine secretions indicates a role in innate host defense. The antibody has been raised against recombinant C-terminally truncated form of CRISP-3 and recognizes both the N-glycosylated and non-glycosylated form of the mature protein.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité