Human recombinant CAR (from Cells)
Fournisseur: ProSci Inc.
91-308
PRSI91-308EA 549 EUR InStock
PRSI91-308
Human recombinant CAR (from Cells)
Protéines et Peptides
Coxsackievirus and Adenovirus Receptor (CAR) belongs to the CTX family of the Ig superfamily. CXADR is a type I transmembrane glycoprotein and expressed in pancreas, brain, heart, small intestine, testis, prostate. It is a receptor that mediates gene transfer and also act as an adhesion molecule within junctional complexes, notably between epithelial cells lining body cavities and within myocardial intercalated discs. CXADR contains an extracellular domain, a transmembrane helix and a C-terminal intracellular domain. The C-terminal interacts with few cytoplasmic junctional proteins, microtubules and the actin cytoskeleton.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité