Human recombinant Cystatin S (from cells)
Fournisseur: ProSci Inc.
91-311
PRSI91-311EA 549 EUR InStock
PRSI91-311
Human recombinant Cystatin S (from cells)
Protéines et Peptides
Cystatin-S, also called Cystatin-SA-III or Salivary acidic protein, is usually expressed in submandibular and sublingual saliva but not in parotid saliva(at protein level). This protein, which is phosphorylated at both its N- and C-terminal regions, belongs to the cystatin family. Cystatin-S can function as an inhibitor to papain or ficin, it partially inhibits stem bromelain and bovine cathepsin C.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité