Human recombinant Cystatin SA (from cells)
Fournisseur: ProSci Inc.
91-312
PRSI91-312EA 852 EUR InStock
PRSI91-312
Human recombinant Cystatin SA (from cells)
Protéines et Peptides
Cystatin SA is a member of family 2 of the Cystatin superfamily. Together with Cystatins S and SN, it is produced by the salivary gland and secreted largely in the submandibular/sublingual saliva. Cystatin SA inhibits Cathepsin L, but may not inhibit Cathepsins B and H. The functions of Cystatin SA are largely unknown. However, it has been suggested to play a role in the control of periodontal tissue destruction.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité