Human recombinant TIM3 (from cells)
Fournisseur: ProSci Inc.
91-327
PRSI91-327EA 528 EUR InStock
PRSI91-327
Human recombinant TIM3 (from cells)
Protéines et Peptides
T-Cell Membrane Protein 3 (TIM3) is a single-pass type I membrane protein that belongs to the TIM family of immunoglobulin superfamily. TIM3 includes a signal sequence (aa 1-21), an extracellular region (aa 22-202) with one Ig-like V-type domain, a transmembrane segment (aa 203-223), and a cytoplasmic domain (aa 224 - 301). TIM3 regulates macrophage activation, inhibits T-helper type 1 lymphocyte (Th1)-mediated auto- and alloimmune responses and promotes immunological tolerance. It may be also involved in T-cell homing and as a receptor for LGALS9.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité