Human recombinant Kallikrein 7 (from cells)
Fournisseur: ProSci Inc.
91-335
PRSI91-335EA 789 EUR InStock
PRSI91-335
Human recombinant Kallikrein 7 (from cells)
Protéines et Peptides
Human Kallikrein 7 is a member of the tissue kallikrein family of extracellular serine proteases that is made up of 15 members. It is predominantly expressed in the skin. A major physiological function of Kallikrein 7 is to regulate the desquamation process (the shedding of corneocytes from the outer layer of the epidermis) through proteolysis of the intercellular adhesive structures between corneocytes. Dysregulation of Kallikrein 7 has been linked to several inflammatory skin diseases including atopic dermatitis, psoriasis, and Netherton syndrome. Studies have shown that Kallikrein 5 is a potential physiological activator for Kallikrein 7. The proform of Kallikrein 7 can be activated by thermolysin.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité